Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114938.2 | NP_001108410.2 | 13 | 2179 | ||
NM_001190182.1 | NP_001177111.1 | 13 | 2152 | |||
XM_011540812.1 | XP_011539114.1 | 11 | 3075 | |||
XM_011540814.1 | XP_011539116.1 | 12 | 3006 | |||
XM_017000450.1 | XP_016855939.1 | 11 | 3048 | |||
XM_017000451.1 | XP_016855940.1 | 10 | 2979 | |||
XM_017000452.1 | XP_016855941.1 | 12 | 2976 | |||
XM_017000453.1 | XP_016855942.1 | 10 | 2967 | |||
XM_017000454.1 | XP_016855943.1 | 11 | 2146 | |||
XM_017000455.1 | XP_016855944.1 | 10 | 1693 | |||
XM_017000456.1 | XP_016855945.1 | 10 | 1694 | |||
GenBank mRNA | AK057646.1 | - | 13 | 2252 | ||
AK298104.1 | - | 13 | 2150 | |||
BC029888.1 | - | 11 | 1802 | 5175485 |