Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195811.1 | NP_001182740.1 | 5 | 635 | ||
NM_001195812.1 | NP_001182741.1 | 4 | 473 | |||
NM_001320272.1 | NP_001307201.1 | 6 | 929 | |||
NM_152666.2 | NP_689879.2 | 6 | 922 | |||
XM_006711752.2 | XP_006711815.1 | 5 | 1089 | |||
XM_011544115.2 | XP_011542417.1 | 7 | 997 | |||
XM_011544116.2 | XP_011542418.1 | 7 | 825 | |||
XM_011544119.2 | XP_011542421.2 | 2 | 996 | |||
XM_011544120.2 | XP_011542422.1 | 5 | 641 | |||
XM_011544121.2 | XP_011542423.1 | 2 | 130 | |||
XM_011544122.2 | XP_011542424.1 | 2 | 1591 | |||
XM_017000567.1 | XP_016856056.1 | 5 | 1102 | |||
XM_017000568.1 | XP_016856057.1 | 5 | 623 | |||
XM_017000569.1 | XP_016856058.1 | 5 | 2602 | |||
XM_017000570.1 | XP_016856059.1 | 3 | 1342 | |||
GenBank mRNA | AK091691.1 | - | 4 | 447 | ||
AK098092.1 | - | 6 | 929 | |||
AK295421.1 | - | 5 | 616 | |||
AK307857.1 | - | 5 | 614 | |||
AY391836.1 | - | 4 | 485 | |||
AY461578.1 | - | 5 | 635 | |||
BC101373.1 | - | 5 | 562 | 40025860 | ||
BC101374.1 | - | 5 | 562 | 40025861 | ||
BC101375.1 | - | 5 | 562 | 40025862 |