Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153367.3 | NP_699198.2 | 3 | 789 | ||
XM_011539452.2 | XP_011537754.1 | 3 | 740 | |||
GenBank mRNA | AK075279.1 | - | 3 | 641 | ||
AK293361.1 | - | 3 | 428 | |||
AK303734.1 | - | 3 | 420 | |||
AK308876.1 | - | 4 | 720 | |||
BC028617.1 | - | 3 | 791 | 4821347 | ||
BC039249.1 | - | 3 | 784 | |||
BC053564.1 | - | 3 | 688 | 6193781 |