Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM71A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153606.3 | NP_705834.2 | 1 | 1766 | ||
GenBank mRNA | AK057358.1 | - | 1 | 1379 | ||
AK301981.1 | - | 2 | 1012 | |||
AK315021.1 | - | 1 | 1390 | |||
BC035007.1 | - | 1 | 1741 | 5267155 |