Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038633.3 | NP_001033722.1 | 8 | 2575 | ||
NM_001242908.1 | NP_001229837.1 | 7 | 2396 | |||
NM_001242909.1 | NP_001229838.1 | 7 | 2075 | |||
NM_001242910.1 | NP_001229839.1 | 6 | 2207 | |||
XM_006710583.3 | XP_006710646.1 | 6 | 2311 | |||
GenBank mRNA | AK098225.1 | - | 7 | 1819 | ||
BC114966.1 | - | 6 | 1995 | 40084277 | ||
DQ165084.1 | - | 7 | 2140 | |||
DQ165085.1 | - | 6 | 1951 |