Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEGR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173808.2 | NP_776169.2 | 5, 6 | 897 | ||
XM_011541200.2 | XP_011539502.1 | 5, 6 | 1153 | |||
GenBank mRNA | AK092307.1 | - | 5, 6 | 804 | ||
AK295481.1 | - | 4, 5 | 663 | |||
BC036771.1 | - | 5, 6 | 819 | 5576828 | ||
BX538014.1 | - | 5, 6 | 801 | |||
BX648027.1 | - | 1, 2 | 28 |