Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXLNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175852.3 | NP_787048.1 | 3 | 446 | ||
XM_011540931.2 | XP_011539233.1 | 3 | 461 | |||
XM_011540932.2 | XP_011539234.1 | 2 | 298 | |||
XM_017000561.1 | XP_016856050.1 | 2 | 739 | |||
XM_017000562.1 | XP_016856051.1 | 2 | 739 | |||
XM_017000563.1 | XP_016856052.1 | 3 | 521 | |||
XM_017000564.1 | XP_016856053.1 | 2 | 300 | |||
XM_017000565.1 | XP_016856054.1 | 2 | 295 | |||
GenBank mRNA | AF516206.1 | - | 2 | 459 | ||
AL832338.1 | - | 3 | 483 | |||
AL832636.2 | - | 3 | 505 | |||
AL832637.2 | - | 2 | 663 | |||
BC080578.1 | - | 2 | 423 | 6598234 | ||
BC103823.1 | - | 2 | 411 | 40001224 | ||
BC103824.1 | - | 2 | 411 | 40001225 |