Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAYN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258390.1 | NP_001245319.1 | 8 | 1857 | ||
NM_001258391.1 | NP_001245320.1 | 6 | 1675 | |||
NM_001318799.1 | NP_001305728.1 | 7 | 1467 | |||
NM_178834.4 | NP_849156.1 | 7 | 1833 | |||
XM_006718769.3 | XP_006718832.1 | 7 | 1446 | |||
XM_006718770.3 | XP_006718833.1 | 6 | 1503 | |||
GenBank mRNA | AK055539.1 | - | 7 | 1834 | ||
AK055654.1 | - | 7 | 1621 | |||
AK296231.1 | - | 6 | 1463 | |||
AY358554.1 | - | 8 | 1682 | |||
BC025407.1 | - | 7 | 1626 | 4820414 |