Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SZT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015284.3 | NP_056099.3 | 2 | 158 | ||
XM_005270686.3 | XP_005270743.1 | 3 | 219 | |||
XM_006710501.3 | XP_006710564.1 | 2 | 106 | |||
XM_011541106.2 | XP_011539408.1 | 2 | 257 | |||
XM_017000819.1 | XP_016856308.1 | 3 | 219 | |||
XM_017000820.1 | XP_016856309.1 | 3 | 219 | |||
GenBank mRNA | AK091821.1 | - | 2 | 79 | ||
BC017576.1 | - | 2 | 162 | |||
BC051343.1 | - | 2 | 92 |