Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF438 SIRNA Assays ›
Gene Symbol:
ZNF438
Gene Name:
zinc finger protein 438
Gene Aliases:
bA330O11.1
Chromosome Location:
Chr.10: 30844634 - 31032406 on Build GRCh38
UniGene ID:
Hs.660642
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
124423
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF438
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001143766.1
NP_001137238.1
7
1650
NM_001143767.1
NP_001137239.1
5
1463
NM_001143768.1
NP_001137240.1
6
1521
NM_001143769.1
NP_001137241.1
7
1775
NM_001143770.1
NP_001137242.1
8
1701
NM_001143771.1
NP_001137243.1
7
1672
NM_182755.2
NP_877432.1
7
1602
NR_026560.1
6
685
XM_006717393.3
XP_006717456.1
5
1417
XM_006717396.3
XP_006717459.1
4
1351
XM_006717398.3
XP_006717461.1
8
1642
XM_006717399.3
XP_006717462.1
7
1575
XM_011519371.2
XP_011517673.1
6
3625
XM_011519372.2
XP_011517674.1
6
1652
XM_011519373.2
XP_011517675.1
5
3567
XM_011519374.2
XP_011517676.1
3
1394
XM_011519376.2
XP_011517678.1
8
1727
XM_011519377.2
XP_011517679.1
8
1723
XM_017015863.1
XP_016871352.1
3
1217
XM_017015864.1
XP_016871353.1
8
3861
XM_017015865.1
XP_016871354.1
7
3802
XM_017015866.1
XP_016871355.1
7
1692
XM_017015867.1
XP_016871356.1
6
3607
XM_017015868.1
XP_016871357.1
5
1913
XM_017015869.1
XP_016871358.1
5
3548
XM_017015870.1
XP_016871359.1
6
2447
XM_017015871.1
XP_016871360.1
5
1907
XM_017015872.1
XP_016871361.1
6
3605
XM_017015873.1
XP_016871362.1
7
3782
XM_017015874.1
XP_016871363.1
5
1468
XM_017015875.1
XP_016871364.1
7
671
XM_017015876.1
XP_016871365.1
9
1420
XM_017015877.1
XP_016871366.1
8
850
XM_017015878.1
XP_016871367.1
7
777
XM_017015879.1
XP_016871368.1
7
857
XM_017015880.1
XP_016871369.1
6
660
XM_017015881.1
XP_016871370.1
5
598
XM_017015882.1
XP_016871371.1
6
705
GenBank mRNA
AF428258.1
-
6
1497
AK057323.1
-
6
673
AK131357.1
-
7
1775
AK292730.1
-
7
1568
AK294253.1
-
8
596
AL833056.1
-
2
1211
BC101622.1
-
3
1227
8069128
BC104757.1
-
3
1227
8143760
BC143462.1
-
4
1326
BC143464.1
-
4
1406
DQ356011.1
-
7
1596
DQ356012.1
-
7
1587
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0