Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBXN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077262.1 | NP_001070730.1 | 6, 7 | 648 | ||
NM_145345.2 | NP_663320.2 | 10, 11 | 1128 | |||
NM_183008.2 | NP_892120.2 | 11, 12 | 1133 | |||
GenBank mRNA | AF520989.1 | - | 9, 10 | 837 | ||
AF521017.1 | - | 9, 10 | 838 | |||
AF521018.1 | - | 9, 10 | 825 | |||
AK093258.1 | - | 11, 12 | 1077 | |||
AK094198.1 | - | 6, 7 | 580 | |||
AK097368.1 | - | 10, 11 | 1226 | |||
AY007163.1 | - | 7, 8 | 592 |