Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SASS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304829.1 | NP_001291758.1 | 2 | 375 | ||
NM_194292.2 | NP_919268.1 | 2 | 375 | |||
XM_017000486.1 | XP_016855975.1 | 2 | 141 | |||
GenBank mRNA | AK302513.1 | - | 3 | 125 | ||
AK307824.1 | - | 2 | 242 | |||
AY359522.1 | - | 2 | 235 | |||
BC054889.1 | - | 2 | 238 | |||
BC101026.3 | - | 2 | 168 | 40007432 | ||
BX641109.1 | - | 2 | 194 | |||
BX648406.1 | - | 2 | 131 |