Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198474.3 | NP_940876.2 | 2 | 676 | ||
XM_005252878.4 | XP_005252935.1 | 3 | 453 | |||
GenBank mRNA | AK298137.1 | - | 3 | 271 | ||
AY358591.1 | - | 2 | 278 | |||
BC035895.1 | - | 2 | 503 | 5229159 | ||
BC092488.1 | - | 2 | 521 | 30531479 | ||
BX647908.1 | - | 2 | 564 | |||
DB257515.1 | - | 2 | 544 |