Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CACNA1C SIRNA Assays ›
Gene Symbol:
CACNA1C
Gene Name:
calcium voltage-gated channel subunit alpha1 C
Gene Aliases:
CACH2, CACN2, CACNL1A1, CCHL1A1, CaV1.2, LQT8, TS
Chromosome Location:
Chr.12: 1969677 - 2697949 on Build GRCh38
UniGene ID:
Hs.118262
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
124625
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CACNA1C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000719.6
NP_000710.5
47
10856
NM_001129827.1
NP_001123299.1
49
11000
NM_001129829.1
NP_001123301.1
47
10979
NM_001129830.2
NP_001123302.2
48
10926
NM_001129831.1
NP_001123303.1
48
10940
NM_001129832.1
NP_001123304.1
48
10916
NM_001129833.1
NP_001123305.1
47
10913
NM_001129834.1
NP_001123306.1
47
10913
NM_001129835.1
NP_001123307.1
47
10913
NM_001129836.1
NP_001123308.1
47
10907
NM_001129837.1
NP_001123309.1
46
10880
NM_001129838.1
NP_001123310.1
46
10880
NM_001129839.1
NP_001123311.1
46
10874
NM_001129840.1
NP_001123312.1
47
10856
NM_001129841.1
NP_001123313.1
47
10856
NM_001129842.1
NP_001123314.1
47
10856
NM_001129843.1
NP_001123315.1
47
10856
NM_001129844.1
NP_001123316.1
47
10847
NM_001129846.1
NP_001123318.1
46
10823
NM_001167623.1
NP_001161095.1
47
10856
NM_001167624.2
NP_001161096.2
48
10926
NM_001167625.1
NP_001161097.1
47
11036
NM_199460.3
NP_955630.3
50
11070
XM_006719017.2
XP_006719080.1
47
10910
XM_011521020.2
XP_011519322.1
48
10985
XM_011521023.2
XP_011519325.1
48
11170
XM_017019926.1
XP_016875415.1
53
12751
XM_017019927.1
XP_016875416.1
52
12646
XM_017019928.1
XP_016875417.1
51
12433
XM_017019929.1
XP_016875418.1
51
12376
XM_017019935.1
XP_016875424.1
49
12172
XM_017019953.1
XP_016875442.1
47
10910
XM_017019954.1
XP_016875443.1
47
10910
XM_017019955.1
XP_016875444.1
49
12172
GenBank mRNA
AK092125.1
-
1
901
AY129012.1
-
1
1895
BC093695.1
-
1
179
Back To Top
More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
immune system development
positive regulation of cytosolic calcium ion concentration
heart development
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
embryonic forelimb morphogenesis
calcium-mediated signaling using extracellular calcium source
camera-type eye development
regulation of insulin secretion
calcium ion transport into cytosol
cardiac conduction
calcium ion transmembrane transport via high voltage-gated calcium channel
calcium ion transmembrane transport
cardiac muscle cell action potential involved in contraction
membrane depolarization during cardiac muscle cell action potential
membrane depolarization during AV node cell action potential
cell communication by electrical coupling involved in cardiac conduction
regulation of heart rate by cardiac conduction
regulation of ventricular cardiac muscle cell action potential
membrane depolarization during atrial cardiac muscle cell action potential
voltage-gated calcium channel activity
protein binding
calmodulin binding
high voltage-gated calcium channel activity
metal ion binding
alpha-actinin binding
voltage-gated calcium channel activity involved in cardiac muscle cell action potential
voltage-gated calcium channel activity involved in AV node cell action potential
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0