Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGFBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005130.4 | NP_005121.1 | 3 | 750 | ||
GenBank mRNA | AK291307.1 | - | 2 | 572 | ||
BC003628.2 | - | 2 | 552 | 3030037 | ||
BC008910.2 | - | 2 | 563 | 3689309 | ||
BC018852.2 | - | 2 | 564 | |||
BQ435589.1 | - | 2 | 633 | |||
M60047.1 | - | 2 | 552 |