Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIOBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039141.2 | NP_001034230.1 | 16 | 6333 | ||
NM_007032.5 | NP_008963.3 | 6 | 927 | |||
NM_138632.2 | NP_619538.2 | 6 | 927 | |||
GenBank mRNA | AF281030.1 | - | 6 | 911 | ||
AL713698.1 | - | 1 | 29 | |||
BC003618.2 | - | 6 | 849 | |||
BC004303.1 | - | 1 | 317 | |||
BC013278.2 | - | 1 | 104 | |||
BC022200.1 | - | 6 | 921 | |||
DQ228003.1 | - | 14 | 5996 | |||
DQ228005.1 | - | 15 | 6198 |