Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HACL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284413.1 | NP_001271342.1 | 15 | 1901 | ||
NM_001284415.1 | NP_001271344.1 | 14 | 1802 | |||
NM_001284416.1 | NP_001271345.1 | 13 | 1736 | |||
NM_012260.3 | NP_036392.2 | 16 | 1982 | |||
NR_104315.1 | 14 | 1803 | ||||
GenBank mRNA | AF161397.1 | - | 16 | 1602 | ||
AJ131753.1 | - | 16 | 1692 | |||
AK056977.1 | - | 8 | 976 | |||
AK301546.1 | - | 15 | 1618 | |||
AK301990.1 | - | 13 | 1471 | |||
AK302086.1 | - | 14 | 1519 | |||
BC001627.1 | - | 16 | 1700 | 3538159 | ||
BC007440.2 | - | 14 | 1508 |