Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040101.1 | NP_001035190.1 | 5 | 1073 | ||
NM_001287763.1 | NP_001274692.1 | 6 | 1128 | |||
NM_001287764.1 | NP_001274693.1 | 5 | 1019 | |||
NM_014392.4 | NP_055207.1 | 5 | 896 | |||
XM_017008022.1 | XP_016863511.1 | 8 | 2313 | |||
GenBank mRNA | AK094573.1 | - | 10 | 2736 | ||
AK125065.1 | - | 1 | 2546 | |||
AK125172.1 | - | 6 | 1110 | |||
AK299657.1 | - | 5 | 681 | |||
AK301911.1 | - | 4 | 637 | |||
AX775767.1 | - | 5 | 820 | |||
BC001745.1 | - | 5 | 820 | 3510777 | ||
BC034227.1 | - | 8 | 1769 | 5265974 |