Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003058.3 | NP_003049.2 | 1 | 389 | ||
GenBank mRNA | AB075951.1 | - | 1 | 393 | ||
AK290787.1 | - | 1 | 293 | |||
BC030978.1 | - | 1 | 389 | |||
BC039899.1 | - | 1 | 270 | 5186229 | ||
X98333.1 | - | 1 | 359 |