Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198763.1 | NP_001185692.1 | 2, 3 | 246 | ||
NM_001198764.1 | NP_001185693.1 | 1, 2 | 135 | |||
NM_014880.4 | NP_055695.2 | 2, 3 | 246 | |||
GenBank mRNA | AF499000.1 | - | 2, 3 | 200 | ||
AK291279.1 | - | 2, 3 | 199 | |||
AK304422.1 | - | 2, 3 | 233 | |||
AY314007.1 | - | 2, 3 | 233 | |||
BC020646.1 | - | 1, 2 | 82 | 4716449 | ||
D14664.1 | - | 2, 3 | 187 | |||
DB217404.1 | - | 2, 3 | 246 | |||
DB460994.1 | - | 1, 2 | 102 |
Gene Symbol: LY75-CD302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198759.1 | NP_001185688.1 | 35, 36 | 5186 | ||
NM_001198760.1 | NP_001185689.1 | 34, 35 | 5018 | |||
GenBank mRNA | AY184222.1 | - | 35, 36 | 5112 | ||
AY314006.1 | - | 34, 35 | 4944 |