Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM19A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082967.2 | NP_001076436.1 | 2, 3 | 402 | ||
NM_015381.6 | NP_056196.2 | 2, 3 | 423 | |||
GenBank mRNA | AF131851.1 | - | 2, 3 | 368 | ||
AK123443.1 | - | 3, 4 | 362 | |||
AK125256.1 | - | 2, 3 | 423 | |||
AY325118.1 | - | 2, 3 | 397 | |||
AY358847.1 | - | 3, 4 | 298 | |||
BC039396.1 | - | 2, 3 | 381 | 5299791 |