Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEPRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319111.1 | NP_001306040.1 | 3 | 290 | ||
NM_001319114.1 | NP_001306043.1 | 3 | 294 | |||
NM_001319115.1 | NP_001306044.1 | 3 | 290 | |||
NM_015412.3 | NP_056227.2 | 4 | 420 | |||
XM_005247276.2 | XP_005247333.1 | 4 | 387 | |||
XM_011512635.2 | XP_011510937.1 | 4 | 383 | |||
XM_011512636.2 | XP_011510938.1 | 4 | 387 | |||
XM_011512638.2 | XP_011510940.1 | 3 | 282 | |||
XM_017006102.1 | XP_016861591.1 | 4 | 383 | |||
XM_017006103.1 | XP_016861592.1 | 3 | 278 | |||
GenBank mRNA | AK024325.1 | - | 4 | 378 | ||
AK300811.1 | - | 3 | 223 | |||
AK301161.1 | - | 3 | 208 | |||
AK304083.1 | - | 3 | 223 | |||
AL117573.1 | - | 4 | 362 | |||
AL537045.3 | - | 3 | 215 | |||
BC014673.2 | - | 4 | 418 | |||
BC067743.1 | - | 4 | 380 | 4812681 | ||
BM152568.1 | - | 3 | 266 |