Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THUMPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114092.1 | NP_001107564.1 | 9 | 1471 | ||
NM_015453.2 | NP_056268.2 | 9 | 1676 | |||
XM_005265021.4 | XP_005265078.1 | 9 | 2033 | |||
XM_005265022.4 | XP_005265079.1 | 9 | 2236 | |||
XM_005265023.3 | XP_005265080.1 | 9 | 1619 | |||
XM_005265024.4 | XP_005265081.1 | 9 | 1619 | |||
XM_006713086.3 | XP_006713149.1 | 9 | 2275 | |||
XM_011533570.2 | XP_011531872.1 | 9 | 1739 | |||
XM_011533571.2 | XP_011531873.1 | 7 | 1490 | |||
XM_017006113.1 | XP_016861602.1 | 8 | 1917 | |||
XM_017006114.1 | XP_016861603.1 | 8 | 2159 | |||
XM_017006115.1 | XP_016861604.1 | 8 | 1560 | |||
XM_017006116.1 | XP_016861605.1 | 8 | 1546 | |||
XM_017006117.1 | XP_016861606.1 | 7 | 1847 | |||
XM_017006118.1 | XP_016861607.1 | 7 | 2089 | |||
XM_017006119.1 | XP_016861608.1 | 7 | 1476 | |||
GenBank mRNA | AK001680.1 | - | 9 | 1435 | ||
AK023263.1 | - | 9 | 1657 | |||
AL117483.1 | - | 6 | 894 | |||
BC001622.1 | - | 9 | 1450 | 3355978 | ||
BC010421.1 | - | 9 | 2007 | 3140059 |