Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNC2LI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193464.1 | NP_001180393.1 | 4, 5 | 340 | ||
NM_015522.3 | NP_056337.1 | 4, 5 | 340 | |||
NM_016008.3 | NP_057092.2 | 4, 5 | 340 | |||
XM_005264364.4 | XP_005264421.1 | 4, 5 | 317 | |||
XM_005264365.4 | XP_005264422.1 | 4, 5 | 317 | |||
GenBank mRNA | AF151818.1 | - | 4, 5 | 301 | ||
AK074206.1 | - | 4, 5 | 330 | |||
AK223432.1 | - | 4, 5 | 340 | |||
AL050006.1 | - | 3, 4 | 193 | |||
AY083346.1 | - | 4, 5 | 300 | |||
BC006969.1 | - | 4, 5 | 299 | 3828551 | ||
BC016324.1 | - | 4, 5 | 334 | 4093010 | ||
BC040558.1 | - | 4, 5 | 315 | |||
BC058823.1 | - | 4, 5 | 307 | 4093493 | ||
BG716986.1 | - | 4, 5 | 345 | |||
BX493698.1 | - | 4, 5 | 343 |