Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YPEL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127399.1 | NP_001120871.1 | 4 | 1378 | ||
NM_001127400.1 | NP_001120872.1 | 4 | 1614 | |||
NM_001127401.1 | NP_001120873.1 | 5 | 1675 | |||
NM_016061.2 | NP_057145.1 | 3 | 1317 | |||
XM_017004318.1 | XP_016859807.1 | 5 | 1913 | |||
XM_017004319.1 | XP_016859808.1 | 4 | 1351 | |||
XM_017004320.1 | XP_016859809.1 | 4 | 1852 | |||
XM_017004321.1 | XP_016859810.1 | 4 | 1544 | |||
XM_017004322.1 | XP_016859811.1 | 4 | 1775 | |||
GenBank mRNA | AF151885.1 | - | 3 | 1239 | ||
AK025730.1 | - | 4 | 1255 |