Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016275.3 | NP_057359.2 | 6 | 830 | ||
GenBank mRNA | AF131856.1 | - | 6 | 785 | ||
AF195141.1 | - | 6 | 788 | |||
AK075006.1 | - | 6 | 803 | |||
AK300315.1 | - | 6 | 818 | |||
AY358095.1 | - | 6 | 794 | |||
AY358096.1 | - | 6 | 798 | |||
BC006012.1 | - | 6 | 821 | 4333083 | ||
BC008411.1 | - | 6 | 788 | 4132578 | ||
BC009556.1 | - | 6 | 762 | 3899430 | ||
BC009611.1 | - | 6 | 782 | 4093231 | ||
BC026350.1 | - | 6 | 810 | 4796883 | ||
BC036738.1 | - | 6 | 781 | 5540243 | ||
BC071699.1 | - | 6 | 773 | 5548565 |