Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003082.3 | NP_003073.1 | 3 | 418 | ||
GenBank mRNA | AK314933.1 | - | 3 | 418 | ||
BC014984.1 | - | 3 | 326 | 4872279 | ||
BC019038.2 | - | 3 | 322 | 4590014 | ||
U44754.1 | - | 3 | 326 | |||
Z47542.1 | - | 3 | 338 |