Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003091.3 | NP_003082.1 | 3 | 353 | ||
NM_198216.1 | NP_937859.1 | 3 | 353 | |||
GenBank mRNA | AK301034.1 | - | 4 | 409 | ||
AK301206.1 | - | 3 | 287 | |||
AK312620.1 | - | 3 | 288 | |||
AY742712.1 | - | 2 | 190 | |||
BC080516.1 | - | 3 | 271 | 2822685 | ||
BM559678.1 | - | 353 | ||||
CR456969.1 | - | 2 | 190 | |||
J04564.1 | - | 3 | 316 | |||
M34081.1 | - | 2 | 169 | |||
M34082.1 | - | 2 | 169 | |||
X15893.1 | - | 2 | 190 | |||
X17567.1 | - | 3 | 244 | |||
X17568.1 | - | 3 | 229 |