Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNMAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103149.1 | NP_001096619.1 | 2 | 986 | ||
NM_018215.3 | NP_060685.2 | 2 | 986 | |||
XM_011527067.2 | XP_011525369.1 | 2 | 1000 | |||
GenBank mRNA | AK096326.1 | - | 2 | 546 | ||
AK290218.1 | - | 3 | 869 | |||
AL834344.1 | - | 2 | 830 | |||
BC026026.1 | - | 1 | 19 | |||
BC032508.1 | - | 2 | 964 | 5246377 | ||
BC051688.1 | - | 2 | 943 | 5295669 | ||
BX649091.1 | - | 1 | 81 |