Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018316.1 | NP_060786.1 | 3 | 2287 | ||
GenBank mRNA | AK001940.1 | - | 3 | 2287 | ||
AK222656.1 | - | 4 | 2207 | |||
AK294074.1 | - | 4 | 848 | |||
BC026319.1 | - | 3 | 2281 | 4799495 |