Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321463.1 | NP_001308392.1 | 13 | 1692 | ||
NM_020170.3 | NP_064555.2 | 13 | 1695 | |||
GenBank mRNA | AK125002.1 | - | 13 | 1751 | ||
AK172848.1 | - | 13 | 1642 | |||
AK314044.1 | - | 13 | 1630 | |||
AL136567.1 | - | 13 | 1526 | |||
AL365369.1 | - | 1 | 38 | |||
BC003076.2 | - | 5 | 382 | |||
BC007275.1 | - | 7 | 671 | |||
BC008920.2 | - | 9 | 902 | |||
BC010064.2 | - | 13 | 1611 | 4100399 | ||
BC013283.2 | - | 13 | 1972 | 3959506 | ||
BC019091.2 | - | 13 | 1611 | 4895124 | ||
BC025926.1 | - | 13 | 1604 | 3509966 | ||
CR533566.1 | - | 13 | 1508 |