Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POGLUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152305.2 | NP_689518.1 | 3 | 329 | ||
NR_024265.1 | 3 | 329 | ||||
XM_006713705.3 | XP_006713768.1 | 2 | 140 | |||
XM_017006878.1 | XP_016862367.1 | 2 | 160 | |||
XM_017006879.1 | XP_016862368.1 | 3 | 275 | |||
XM_017006880.1 | XP_016862369.1 | 3 | 275 | |||
GenBank mRNA | AF168711.1 | - | 3 | 280 | ||
AK075444.1 | - | 3 | 265 | |||
AK222937.1 | - | 3 | 289 | |||
AK295984.1 | - | 3 | 304 | |||
AK301002.1 | - | 3 | 304 | |||
AK315367.1 | - | 3 | 304 | |||
AY298903.1 | - | 3 | 315 | |||
AY358581.1 | - | 3 | 272 | |||
BC030614.2 | - | 3 | 315 | 4796951 | ||
BC048810.1 | - | 3 | 273 | 5243487 | ||
BP347179.1 | - | 3 | 329 | |||
CD517010.1 | - | 3 | 245 |