Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALGAPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282917.1 | NP_001269846.1 | 13 | 2097 | ||
NM_001282918.1 | NP_001269847.1 | 13 | 2239 | |||
NM_020336.3 | NP_065069.1 | 13 | 2239 | |||
XM_005260462.2 | XP_005260519.1 | 14 | 2233 | |||
XM_005260463.2 | XP_005260520.1 | 14 | 2233 | |||
XM_005260464.2 | XP_005260521.1 | 14 | 2233 | |||
XM_005260465.2 | XP_005260522.1 | 13 | 2197 | |||
XM_017027966.1 | XP_016883455.1 | 14 | 2233 | |||
XM_017027967.1 | XP_016883456.1 | 13 | 2197 | |||
XM_017027968.1 | XP_016883457.1 | 13 | 1759 | |||
XM_017027969.1 | XP_016883458.1 | 14 | 2262 | |||
GenBank mRNA | AB033045.3 | - | 13 | 2229 | ||
AK123576.1 | - | 12 | 1898 | |||
AL834436.1 | - | 11 | 1461 | |||
BC047482.1 | - | 13 | 2202 | 5270777 | ||
CR936800.1 | - | 13 | 2003 |