Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304389.1 | NP_001291318.1 | 3 | 1176 | ||
NM_020678.3 | NP_065729.1 | 3 | 1185 | |||
GenBank mRNA | AF225421.1 | - | 3 | 1156 | ||
BC040732.1 | - | 3 | 1176 | 5223743 | ||
BC074741.2 | - | 3 | 1046 | 30915199 |