Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278712.1 | NP_001265641.1 | 6 | 795 | ||
NM_022488.4 | NP_071933.2 | 6 | 795 | |||
XM_011513074.1 | XP_011511376.1 | 6 | 405 | |||
GenBank mRNA | AB079384.1 | - | 6 | 360 | ||
AF202092.1 | - | 6 | 375 | |||
AK025778.1 | - | 11 | ||||
AK307432.1 | - | 6 | 345 | |||
AK308919.1 | - | 6 | 756 | |||
AL137515.1 | - | 6 | 495 | |||
BC002830.1 | - | 6 | 753 | 3633903 | ||
BC024221.2 | - | 6 | 470 | 3938447 |