Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022492.4 | NP_071937.4 | 4 | 293 | ||
NR_027749.1 | 3 | 190 | ||||
XM_005264492.1 | XP_005264549.1 | 4 | 282 | |||
XM_011533035.1 | XP_011531337.1 | 4 | 282 | |||
XM_011533036.1 | XP_011531338.1 | 4 | 282 | |||
XM_011533037.1 | XP_011531339.1 | 4 | 282 | |||
XM_011533040.1 | XP_011531342.1 | 3 | 231 | |||
XM_017004706.1 | XP_016860195.1 | 4 | 282 | |||
XM_017004707.1 | XP_016860196.1 | 4 | 282 | |||
XM_017004708.1 | XP_016860197.1 | 3 | 187 | |||
XM_017004709.1 | XP_016860198.1 | 3 | 417 | |||
GenBank mRNA | AK022850.1 | - | 4 | 279 | ||
AK090520.1 | - | 3 | 332 | |||
AK223355.1 | - | 4 | 277 | |||
AK225626.1 | - | 4 | 279 | |||
AK303105.1 | - | 3 | 176 | |||
AK303245.1 | - | 4 | 279 | |||
BC036017.1 | - | 4 | 277 | 5270575 | ||
BC045536.1 | - | 3 | 181 | |||
BC096709.1 | - | 4 | 279 | 40023059 | ||
BC098365.1 | - | 4 | 279 | 40023060 | ||
BC099729.1 | - | 3 | 176 | |||
BX648131.1 | - | 3 | 175 |