Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022492.4 | NP_071937.4 | 13 | 2577 | ||
NR_027749.1 | 12 | 2468 | ||||
XM_005264492.1 | XP_005264549.1 | 13 | 2560 | |||
XM_011533035.1 | XP_011531337.1 | 13 | 2626 | |||
XM_011533036.1 | XP_011531338.1 | 13 | 2623 | |||
XM_011533037.1 | XP_011531339.1 | 13 | 2620 | |||
XM_011533040.1 | XP_011531342.1 | 12 | 2575 | |||
XM_017004706.1 | XP_016860195.1 | 13 | 2563 | |||
XM_017004707.1 | XP_016860196.1 | 13 | 2557 | |||
XM_017004708.1 | XP_016860197.1 | 12 | 2471 | |||
XM_017004709.1 | XP_016860198.1 | 12 | 2695 | |||
GenBank mRNA | AK022850.1 | - | 9 | 2087 | ||
AK090520.1 | - | 8 | 1753 | |||
AK223355.1 | - | 9 | 1265 | |||
AK225626.1 | - | 8 | 1157 | |||
BC045536.1 | - | 12 | 2459 | |||
BX648131.1 | - | 7 | 3513 |