Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022903.3 | NP_075054.3 | 1804 | |||
GenBank mRNA | BC035516.1 | - | 1773 | 5229681 | ||
BC071960.1 | - | 963 |