Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023008.3 | NP_075384.3 | 3, 4 | 309 | ||
XM_011528190.2 | XP_011526492.1 | 2, 3 | 240 | |||
GenBank mRNA | AK023011.1 | - | 1, 2 | 87 | ||
AK093879.1 | - | 2, 3 | 421 | |||
AK300780.1 | - | 3, 4 | 293 | |||
AK300850.1 | - | 3, 4 | 314 | |||
AL833189.1 | - | 2, 3 | 962 | |||
AU280109.1 | - | 3, 4 | 310 | |||
DA438802.1 | - | 3, 4 | 310 |