Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319152.1 | NP_001306081.1 | 2 | 910 | ||
NM_001319153.1 | NP_001306082.1 | 2 | 912 | |||
NM_024059.3 | NP_076964.1 | 2 | 928 | |||
XM_011529041.2 | XP_011527343.1 | 2 | 1465 | |||
XM_017028060.1 | XP_016883549.1 | 2 | 1064 | |||
GenBank mRNA | AK125168.1 | - | 1 | 2785 | ||
BC000912.2 | - | 2 | 928 | 3048411 |