Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171948.1 | NP_001165419.1 | 6 | 1234 | ||
NM_001171949.1 | NP_001165420.1 | 5 | 1138 | |||
NM_024069.3 | NP_076974.2 | 5 | 1103 | |||
XM_006722883.2 | XP_006722946.1 | 6 | 953 | |||
XM_011528272.2 | XP_011526574.1 | 5 | 1809 | |||
XM_011528273.2 | XP_011526575.1 | 5 | 1308 | |||
XM_017027281.1 | XP_016882770.1 | 6 | 1251 | |||
GenBank mRNA | AF173381.1 | - | 6 | 924 | ||
AK091145.1 | - | 5 | 2393 | |||
AK098346.1 | - | 5 | 1077 | |||
BC001080.2 | - | 5 | 1071 | 2823374 | ||
BC006479.1 | - | 5 | 1071 | 2823374 |