Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024334.2 | NP_077310.1 | 12 | 2142 | ||
XM_017007176.1 | XP_016862665.1 | 13 | 2264 | |||
GenBank mRNA | AF086408.1 | - | 1 | 134 | ||
AK027466.1 | - | 6 | 1372 | |||
AK027757.1 | - | 12 | 2068 | |||
AK027827.1 | - | 7 | 1423 | |||
AK027877.1 | - | 12 | 2006 | |||
AK074073.1 | - | 10 | 4573 | |||
AK075010.1 | - | 12 | 1993 | |||
AK225589.1 | - | 12 | 2065 | |||
AL136916.1 | - | 12 | 2035 | |||
BC003125.1 | - | 12 | 2037 | 3503636 | ||
BC008054.2 | - | 10 | 1632 | |||
BC011719.2 | - | 12 | 2037 | 4079766 |