Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP13A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024524.3 | NP_078800.3 | 16 | 2151 | ||
XM_005269357.3 | XP_005269414.1 | 16 | 2172 | |||
XM_011513120.2 | XP_011511422.1 | 17 | 2273 | |||
XM_011513121.1 | XP_011511423.1 | 16 | 2151 | |||
XM_011513122.1 | XP_011511424.1 | 16 | 1845 | |||
XM_011513123.2 | XP_011511425.1 | 17 | 2278 | |||
XM_011513124.2 | XP_011511426.1 | 17 | 2278 | |||
GenBank mRNA | AJ306929.1 | - | 7 | 2675 | ||
AK024639.1 | - | 5 | 548 | |||
AK225064.1 | - | 5 | 548 |