Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287527.1 | NP_001274456.1 | 16 | 4258 | ||
NM_024692.5 | NP_078968.3 | 16 | 4262 | |||
XM_005264562.2 | XP_005264619.1 | 16 | 4289 | |||
XM_005264563.1 | XP_005264620.1 | 16 | 4294 | |||
XM_006712104.2 | XP_006712167.1 | 16 | 4334 | |||
XM_006712105.3 | XP_006712168.1 | 15 | 4103 | |||
XM_011533109.1 | XP_011531411.1 | 16 | 4042 | |||
XM_011533110.1 | XP_011531412.1 | 15 | 4095 | |||
XM_017004958.1 | XP_016860447.1 | 16 | 4121 | |||
GenBank mRNA | AB209042.1 | - | 16 | 5928 | ||
AK023505.1 | - | 1 | 1790 | |||
AL834443.1 | - | 16 | 4237 |