Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMKMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024766.4 | NP_079042.1 | 7, 8 | 732 | ||
XM_011533111.2 | XP_011531413.1 | 7, 8 | 698 | |||
XM_011533113.2 | XP_011531415.1 | 5, 6 | 408 | |||
XM_017004982.1 | XP_016860471.1 | 5, 6 | 2835 | |||
XM_017004983.1 | XP_016860472.1 | 6, 7 | 947 | |||
GenBank mRNA | AK027104.1 | - | 7, 8 | 672 |