Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321154.1 | NP_001308083.1 | 3, 4 | 478 | ||
NM_001321155.1 | NP_001308084.1 | 4, 5 | 602 | |||
NM_001321156.1 | NP_001308085.1 | 4, 5 | 672 | |||
NM_001321157.1 | NP_001308086.1 | 4, 5 | 505 | |||
NM_001321158.1 | NP_001308087.1 | 2, 3 | 226 | |||
NM_001321159.1 | NP_001308088.1 | 2, 3 | 323 | |||
NM_001321160.1 | NP_001308089.1 | 4, 5 | 602 | |||
NM_001321161.1 | NP_001308090.1 | 2, 3 | 226 | |||
NM_001321162.1 | NP_001308091.1 | 3, 4 | 447 | |||
NM_024770.4 | NP_079046.2 | 3, 4 | 381 | |||
XM_011511875.2 | XP_011510177.1 | 4, 5 | 482 | |||
XM_017004984.1 | XP_016860473.1 | 4, 5 | 482 | |||
XM_017004985.1 | XP_016860474.1 | 4, 5 | 580 | |||
XM_017004986.1 | XP_016860475.1 | 3, 4 | 358 | |||
XM_017004987.1 | XP_016860476.1 | 2, 3 | 220 | |||
XM_017004988.1 | XP_016860477.1 | 2, 3 | 204 | |||
XM_017004989.1 | XP_016860478.1 | 3, 4 | 373 | |||
GenBank mRNA | AK024046.1 | - | 3, 4 | 448 | ||
AK124092.1 | - | 4, 5 | 553 | |||
AK292108.1 | - | 3, 4 | 350 | |||
AK297141.1 | - | 2, 3 | 226 | |||
BC025250.1 | - | 4, 5 | 502 | 5433692 | ||
DC349499.1 | - | 2, 3 | 226 |