Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115116.1 | NP_001108588.1 | 3 | 721 | ||
NM_024933.3 | NP_079209.3 | 3 | 721 | |||
XM_005264569.4 | XP_005264626.1 | 3 | 1881 | |||
XM_011533114.2 | XP_011531416.1 | 3 | 557 | |||
GenBank mRNA | AK022118.1 | - | 1 | 1132 | ||
AK093479.1 | - | 3 | 557 | |||
BC035234.1 | - | 3 | 721 | 5166484 | ||
DB036445.1 | - | 2 | 319 |