Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171937.1 | NP_001165408.1 | 6, 7 | 874 | ||
NM_025129.4 | NP_079405.2 | 7, 8 | 982 | |||
NR_033269.1 | 6, 7 | 897 | ||||
XM_006723399.3 | XP_006723462.1 | 7, 8 | 960 | |||
XM_011527339.1 | XP_011525641.1 | 7, 8 | 960 | |||
XM_011527340.1 | XP_011525642.1 | 7, 8 | 822 | |||
XM_011527341.2 | XP_011525643.1 | 7, 8 | 886 | |||
XM_011527342.1 | XP_011525644.1 | 7, 8 | 1109 | |||
XM_011527343.1 | XP_011525645.1 | 7, 8 | 960 | |||
XM_011527345.1 | XP_011525647.1 | 7, 8 | 918 | |||
XM_011527346.1 | XP_011525648.1 | 6, 7 | 836 | |||
XM_011527347.1 | XP_011525649.1 | 8, 9 | 876 | |||
XM_017027319.1 | XP_016882808.1 | 7, 8 | 822 | |||
XM_017027320.1 | XP_016882809.1 | 7, 8 | 960 | |||
XM_017027321.1 | XP_016882810.1 | 6, 7 | 836 | |||
XM_017027322.1 | XP_016882811.1 | 7, 8 | 822 | |||
GenBank mRNA | AK026341.1 | - | 7, 8 | 956 | ||
AK057964.1 | - | 6, 7 | 777 | |||
AK315445.1 | - | 7, 8 | 960 | |||
BC004445.1 | - | 7, 8 | 883 | 2819805 | ||
BC010092.2 | - | 6, 7 | 739 | 3354417 | ||
BC016793.2 | - | 7, 8 | 883 | 2819805 |