Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAA50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308445.1 | NP_001295374.1 | 5 | 702 | ||
NM_025146.3 | NP_079422.1 | 5 | 705 | |||
GenBank mRNA | AK022540.1 | - | 11 | |||
AK023090.1 | - | 5 | 620 | |||
AK023256.1 | - | 5 | 655 | |||
AK296456.1 | - | 5 | 400 | |||
AK307184.1 | - | 6 | 549 | |||
AK315876.1 | - | 5 | 507 | |||
AL601126.1 | - | 5 | 600 | |||
BC012731.2 | - | 5 | 644 | 3619903 |