Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRABD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320484.1 | NP_001307413.1 | 5 | 425 | ||
NM_001320485.1 | NP_001307414.1 | 5 | 425 | |||
NM_001320487.1 | NP_001307416.1 | 5 | 459 | |||
NM_001320488.1 | NP_001307417.1 | 5 | 459 | |||
NM_025204.3 | NP_079480.2 | 5 | 450 | |||
NR_135275.1 | 5 | 425 | ||||
XM_006724422.3 | XP_006724485.1 | 5 | 429 | |||
XM_006724423.3 | XP_006724486.1 | 5 | 422 | |||
XM_011530712.2 | XP_011529014.2 | 5 | 485 | |||
XM_011530713.2 | XP_011529015.2 | 5 | 483 | |||
XM_011530715.2 | XP_011529017.2 | 5 | 488 | |||
XM_011530716.2 | XP_011529018.1 | 5 | 448 | |||
XM_011530717.2 | XP_011529019.1 | 5 | 429 | |||
XM_011530718.2 | XP_011529020.1 | 5 | 408 | |||
XM_017028943.1 | XP_016884432.1 | 5 | 490 | |||
XM_017028944.1 | XP_016884433.1 | 5 | 537 | |||
GenBank mRNA | AK024447.1 | - | 1 | 838 | ||
AK292050.1 | - | 5 | 429 | |||
AL449244.1 | - | 5 | 431 | |||
AY203945.1 | - | 5 | 417 | |||
BC012445.1 | - | 5 | 269 | 3857552 | ||
BC093029.1 | - | 5 | 444 | 30527560 | ||
BI907868.1 | - | 5 | 407 | |||
BU902575.1 | - | 5 | 425 | |||
BX417817.2 | - | 5 | 410 | |||
BX425224.2 | - | 5 | 410 | |||
CF272467.1 | - | 5 | 419 |